Learn More
Eppendorf™ Adapter for Rotor F-35-6-30
Adapter for Rotors designed for use with Eppendorf Centrifuges
Brand:  Eppendorf™ 5427738002
 View more versions of this product
Product Code. 10259013
  • 104.42 € / Pack of 2
Explore more special offers
Description
Description
  • Rotor F-35-6-30 holds 6 x 15/50mL conical or 12 × Vacutainer™ tubes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Large Rotor Bore Adapter
1 x 9 to 15 mL Tube
Rotor F-35-6-30
17 dia. x 90 to 125mm
Round Bottom, Blood Collection Tubes
17 mm
Large rotor bore
2/Pk.