missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Adapter for Centrifuge 5910 R with Rotor S-4xUniversal
Adapter for 1 x 1000 mL bottle in Centrifuge 5910 R with Rotor S-4xUniversal
Brand: Eppendorf™ 5910756003
This item is not returnable.
View return policy
Description
- For 1,000 mL wide-neck bottle (Catalog No. 5910 770.006) in combination with Rotor S-4xUniversal
- Set of 2 pieces
Specifications
1 x 1000 mL | |
For Centrifuge 5910 R with Rotor S-4xUniversal | |
1 | |
Set of 2 pieces |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction