missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Adapter for Conical Tubes

Designed for use with a variety of products to expand liquid handling capabilities in the lab

93.25 € - 263.00 €

Specifications

Type Adapter
Tube Type Conical
Capacity 15 mL
Color Orange
View More Specs

Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10616640
View Documents
Eppendorf™
5820718005
Rotor A-4-81 and Rotor FA-45-6-30
263.00 €
Pack of 7
Please to purchase this item. Need a web account? Register with us today!
10725642
View Documents
Eppendorf™
5820717009
Rotor FA-45-6-30
93.25 €
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Available in packs of two or seven
  • For use with Eppendorf Rotors
Specifications

Specifications

Adapter
Conical
15 mL
Orange
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.