Learn More
Eppendorf™ Adapter S-4x Universal Rotor Large
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4xUniversal-Large rotor.
Brand:  Eppendorf™ 5920748009
335.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15798399
  • 336.34 € / Pack of 2
Explore more special offers
Description
Description
  • Suitable for S-4xUniversal-Large rotor buckets
  • Accommodates selected array of tubes, bottles, or cards
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Conical, Skirted
31 mm
S-4xUniversal-Large
7
7 x 50 mL Conical, Skirted Tubes
141 mm
7
2/Pk.