Learn More
Eppendorf™ Adapter for S-4x400 Bucket
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4x400 rotor.
Brand:  Eppendorf™ 5910702000
 View more versions of this product
Product Code. 15823222
  • 277.00 € / Pack of 2
Explore more special offers
Description
Description
  • Suitable for S-4x400 Rotor buckets
  • Accommodates selected array of tubes or bottles
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Microtube, Conical
17 mm
S-4x400 Bucket
7
7 x 5.0 mL Microtubes, 15 mL Conical Tubes
133 mm
7
2/Pk.