Learn More
Eppendorf™ Adapter for S-4x400 Bucket
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4x400 rotor.
Brand:  Eppendorf™ 5910714009
 View more versions of this product
Product Code. 15823232
  • 283.13 € / Pack of 2
Explore more special offers
Description
Description
  • Suitable for S-4x400 Rotor buckets
  • Accommodates selected array of tubes or bottles
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Conical
62 mm
S-4x400 Bucket
1
1 x 175 to 225 mL Conical Tubes
143 mm
1
2/Pk.