Learn More
Eppendorf™ Adapter for S-4x400 Bucket
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4x400 rotor.
Brand:  Eppendorf™ 5910704003
 View more versions of this product
Product Code. 15843222
  • 283.13 € / Pack of 2
Explore more special offers
Description
Description
  • Suitable for S-4x400 Rotor buckets
  • Accommodates selected array of tubes or bottles
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Round Bottom
16 mm
S-4x400 Bucket
11
11 x 7.5 to 12 mL Round Bottom Tubes
122 mm
11
2/Pk.