Learn More
Eppendorf™ Adapter for S-4x400 Bucket
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4x400 rotor.
Brand: Eppendorf™ 5910706006
167.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Suitable for S-4x400 Rotor buckets
- Accommodates selected array of tubes or bottles
Specifications
1 x 400 mL Bottle | |
1 | |
2/Pk. |
S-4x400 Bucket | |
1 |