Learn More
Eppendorf™ Adapter for S-4xUniversal Rotor
The Adapter for S-4xUniversal Rotor facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for the S-4xUniversal Rotor.
Brand:  Eppendorf™ 5910751001
 View more versions of this product
Product Code. 15820927
  • 472.88 € / Pack of 2
Explore more special offers
Description
Description
  • Suitable for S-4xUniversal rotor buckets
  • Accommodates selected array of tubes, bottles, or plates
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Flat/Conical Bottom
32 mm
S-4xUniversal Rotor
2
7 x 50 mL Conical Tubes, 1 x 96/384-well Microplate, 1 x 250 mL Flat-bottom Bottle, 1 x 175 to 225 mL Conical Bottle
124 mm, 146 mm
7/1/1/1