Learn More
Eppendorf™ Adapter for S-4xUniversal Rotor
The Adapter for S-4xUniversal Rotor facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for the S-4xUniversal Rotor.
Brand:  Eppendorf™ 5910753004
319.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 15840927
  • 320.28 € / Pack of 2
Explore more special offers
Description
Description
  • Suitable for S-4xUniversal rotor buckets
  • Accommodates selected array of tubes, bottles, or plates
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PCR
88 mm
5
5 x MTP/DWP/PCR Plates
S-4xUniversal Rotor
2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Eppendorf™ Adapter for S-4xUniversal Rotor > Capacity: 5 x MTP/DWP/PCR Plates; Tube Type: PCR; No. of Places: 5