Learn More
Eppendorf™ Adapter for S-4xUniversal Rotor
The Adapter for S-4xUniversal Rotor facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for the S-4xUniversal Rotor.
Brand:  Eppendorf™ 5910755007
 View more versions of this product
Product Code. 15860927
  • 320.28 € / Pack of 2
Explore more special offers
Description
Description
  • Suitable for S-4xUniversal rotor buckets
  • Accommodates selected array of tubes, bottles, or plates
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Round Bottom
13 mm
S-4xUniversal Rotor
2
30 x 4 to 8 mL Round Bottom Tubes
119 mm
30