Learn More
Eppendorf™ Adapter for S-4xUniversal Rotor
The Adapter for S-4xUniversal Rotor facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for the S-4xUniversal Rotor.
Brand:  Eppendorf™ 5910758006
 View more versions of this product
Product Code. 15880927
  • 336.34 € / Pack of 2
Explore more special offers
Description
Description
  • Suitable for S-4xUniversal rotor buckets
  • Accommodates selected array of tubes, bottles, or plates
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Round Bottom (FACS)
S-4xUniversal Rotor
2
46 tubes of 12 x 75mm or 5ml, Round Bottom Tubes (FACS)
46