Learn More
HUBER Adapters
Accessories
Brand:  HUBER 6472
 View more versions of this product
Product Code. 10245432
  • 175.00 € / Each
Explore more special offers
Specifications
M30 x 1.5 female thread to 3/4 NPT male thread
1 Ea.
0.2 kg
Adapter