missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Adapters for Fisherbrand™ accuSpin™ 4-Place Rotor with Rectangular Bucket

Product Code. p-2365119 Shop All Thermo Scientific Products
Click to view available options
Container Type:
Bottle
Color:
Brown
Gray
White
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10612422

Brand: Fisherbrand™ 75005327

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Adapters are sold individually and are available in other sizes for convience.

Fit in rectangular bucket 13-100-587 for swinging bucket rotor 13-100-584; also can be used in sealed rotor 13-100-589.

Order Info

Adapters are sold individually. Order correct quantity to outfit rotor. Other adapter sizes are available online.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) accuSpin 1/1R 4-Place Rotor
Color White
Container Type Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.