Learn More
Eppendorf™ Adapters For 100mL Rectangular Bucket
For use with Eppendorf Tubes
Brand:  Eppendorf™ 5804753003
 View more versions of this product
Product Code. 10247251
  • 165.00 € / Pack of 2
Explore more special offers
Description
Description
  • Adapters for rectangular buckets
  • For use with Rotor A-4-44
  • Available in a variety of sizes for a wide range of applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Adapter
100mL Rectangular Bucket
28
2 Pack
3 to 15 mL
16 mm
Standard, Vacutainer, Falcon and micro test tubes