Learn More
Thermo Scientific™ Adapters for TX-1000 Swinging Bucket Rotor
An assortment of adapters for outstanding flexibility
Brand:  Thermo Scientific™ 75004252
1031.13 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
 View more versions of this product
Product Code. 15302127
  • 1085.40 € / Pack of 4
Explore more special offers
Description
Description
An assortment of adapters for outstanding flexibility
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor Adapter
48 x 50 mL
120 mm
50 mL Nalgene Oak Ridge Tube
29.5 mm
50mL Thermo Scientific Nalgene Oak Ridge Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Adapters for TX-1000 Swinging Bucket Rotor > 50mL Thermo Scientific Nalgene Oak Ridge Tube