Learn More
Interscience Adaptor for Petri Dish
Adaptor for Petri dish (55 mm)
Brand:  Interscience 436005
Product Code. 13425199
  • 139.00 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Adaptor for Petri dishes of dia. 55 mm for Scan 300/500/1200 (optional accessory)
Specifications
Specifications
Adaptor for Petri dishes of dia. 55 mm for Scan 300/500/1200 (optional accessory)
Contact Petri Dish
Automatic colony counter, Scan 300/500/1200