Learn More
Cytiva ADCF-MAb Media
Increase process yields for the manufacture of monoclonal antibodies (MAbs) with HyClone™ ADCF-MAb Media, a protein-free media containing no animal-derived components.
Brand:  Cytiva SH30349.01
 View more versions of this product
Product Code. 10788134
  • 44.10 € / 500mL
Explore more special offers
Description
Description
  • ADCF
  • Metabolically designed for high cell yield and monoclonal antibody production
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
  • Contains L-glutamine and Poloxamer 188
  • Formulations without L-glutamine support the GS gene expression system
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Liquid
0.2 μm Sterile-filtered
No Protein, No Phenol Red
cGMP
with L-Glutamine
ADCF-mAb Liquid Medium
Glutamine, Poloxamer 188, HEPES, Sodium Bicarbonate
500 mL
2° to 8°C, away from light
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva ADCF-MAb Media > ADCF MAb; With L Glutamine; 500mL