Learn More
Memmert™ Additional Stainless Steel Grid for SF160, Reinforced Chamber Option
Product Code. 17568135 Shop All Memmert Products
Click to view available options
For Use With (Equipment):
SF160 Sterilizer
Unit Size:
Each
Product Code. 17568135
Brand: Memmert™ SF160E29767
Only null left
Add to basket
Add to basket
Additional grid features a sturdy stainless steel construction.
  • Reinforced
  • Stainless Steel
  • Electropoloished
  • Max. loading 60 kg
Specifications
For Use With (Equipment) SF160 Sterilizer
Product Type Reinforced Grid
Material Stainless Steel