Learn More
Memmert™ Additional Stainless Steel Grid for SN160plus, Reinforced Chamber Option
Additional grid features a sturdy stainless steel construction.
Brand:  Memmert™ SN160plusE29767
Product Code. 17538525
  • 298.00 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Reinforced
  • Stainless Steel
  • Electropoloished
  • Max. loading 60 kg
Specifications
Specifications
SN160plus Sterilizer
Reinforced Grid
Stainless Steel