Learn More
Heidolph Instruments Multi-tier Attachment
For use with Unimax 2010, Promax 2020 and Polymax 2040
Brand:  Heidolph Instruments 549-62000-00
Product Code. 10345632
  • 364.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Includes perforated platform 2000 and 4 supporting rods
Specifications
Specifications
0 to 1100 rpm
25 to 250 mL Erlenmeyer Flasks
Support Floor