missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adenine hydrochloride, 98+%, cont. up to ca 5% water

Product Code. 11408257
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2922-28-3
Molecular Formula C5H6ClN5
Molecular Weight (g/mol) 171.59
MDL Number MFCD00038990
InChI Key UQVDQSWZQXDUJB-UHFFFAOYSA-N
Synonym adenine hydrochloride, 7h-purin-6-amine hydrochloride, 6-aminopurine hydrochloride, 1h-purin-6-amine, monohydrochloride, adenine monohydrochloride, unii-364h11m7od, adenine hcl, 1h-purin-6-amine, hydrochloride, 9h-purin-6-amine, hydrochloride 1:?, 9h-purin-6-amine, hydrochloride 1:1
PubChem CID 76219
IUPAC Name 7H-purin-6-amine;hydrochloride
SMILES Cl.NC1=C2NC=NC2=NC=N1

Specifications

Melting Point ∼290°C (decomposition)
Quantity 5 g
Beilstein 4009585
Formula Weight 171.59
Percent Purity ≥98%
Assay cont. up to ≈5% water
Chemical Name or Material Adenine hydrochloride
compliance-icons
Product Identifier
  • Adenine hydrochloride hemihydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : 1H-Purin-6-amine, monohydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.