missing translation for 'onlineSavingsMsg'
Learn More

Adenine Sulfate Dihydrate, >97%, Thermo Scientific Chemicals

Product Code. 15855048
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Spezifikation

CAS 321-30-2
Melting Point ∼210°C (decomposition)
Packaging Plastic bottle
Molecular Formula C10H12N10O4S
MDL Number MFCD00213655
Quantity 100 g
Beilstein 3820263
Synonym adenine sulfate, adenine hemisulfate, 7 h-purin-6-amine sulfate 2:1, adeninsulfat, adeninium sulfate, adenine sulfate 2:1, adeninsulfat german, 1h-purin-6-amine, sulfate 2:1, diadenine sulphate, unii-741gjf3k9m
InChI Key LQXHSCOPYJCOMD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O.NC1=C2NC=NC2=NC=N1.NC1=C2NC=NC2=NC=N1
IUPAC Name 7 H-purin-6-amine;sulfuric acid
Molecular Weight (g/mol) 368.33
PubChem CID 9449
Formula Weight 404.36
Percent Purity ≥97%
Physical Form Crystalline Powder
Chemical Name or Material Adenine Sulfate Dihydrate
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'