Learn More
Adenosine-2',3'-cyclic monophosphate sodium salt, 97%, Thermo Scientific Chemicals
175.00 € - 725.00 €
Chemical Identifiers
CAS | 37063-35-7 |
---|---|
Molecular Formula | C10H11N5NaO6P |
Molecular Weight (g/mol) | 351.19 |
MDL Number | MFCD00005757 |
InChI Key | VSDSIACSNXHGOV-GWKNMROSNA-M |
Synonym | adenosine-2 inverted exclamation mark ,3 inverted exclamation mark-cyclic monophosphate sodium salt |
PubChem CID | 131698406 |
IUPAC Name | [(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2-oxido-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3,2]dioxaphosphol-6-yl]methanol;sodium |
SMILES | [Na+].NC1=C2N=CN([C@@H]3O[C@H](CO)[C@H]4OP([O-])(=O)O[C@@H]34)C2=NC=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15470137
|
Thermo Scientific Alfa Aesar
J60558.MC |
100 mg |
175.00 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15480137
|
Thermo Scientific Alfa Aesar
J60558.MD |
250 mg |
373.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15490137
|
Thermo Scientific Alfa Aesar
J60558.ME |
500 mg | |||||||
Description
Adenosine-2’,3’-cyclic Monophosphate is an Adenosine (A280400) derivative useful as anticancer agent. It is also used as a Pharmaceutical intermediate, pharmaceutical research and chemical analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAdenosine-2’,3’-cyclic Monophosphate is an Adenosine (A280400) derivative useful as anticancer agent. It is also used as a Pharmaceutical intermediate, pharmaceutical research and chemical analysis.
Solubility
It is soluble in DMSO, water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic. Keep away fom strong oxidizing agents.
Chemical Identifiers
37063-35-7 | |
351.19 | |
VSDSIACSNXHGOV-GWKNMROSNA-M | |
131698406 | |
[Na+].NC1=C2N=CN([C@@H]3O[C@H](CO)[C@H]4OP([O-])(=O)O[C@@H]34)C2=NC=N1 |
C10H11N5NaO6P | |
MFCD00005757 | |
adenosine-2 inverted exclamation mark ,3 inverted exclamation mark-cyclic monophosphate sodium salt | |
[(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2-oxido-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3,2]dioxaphosphol-6-yl]methanol;sodium |
Specifications
37063-35-7 | |
White | |
MFCD00005757 | |
It is soluble in DMSO,water. | |
[Na+].NC1=C2N=CN([C@@H]3O[C@H](CO)[C@H]4OP([O-])(=O)O[C@@H]34)C2=NC=N1 | |
351.19 | |
351.19 | |
Powder |
241°C to 243°C | |
C10H11N5NaO6P | |
adenosine-2 inverted exclamation mark ,3 inverted exclamation mark-cyclic monophosphate sodium salt | |
VSDSIACSNXHGOV-GWKNMROSNA-M | |
[(3aR,4R,6R,6aR)-4-(6-aminopurin-9-yl)-2-oxido-2-oxo-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3,2]dioxaphosphol-6-yl]methanol;sodium | |
131698406 | |
97% | |
Adenosine-2',3'-cyclic monophosphate sodium salt |
Safety and Handling
missing translation for 'einecsNumber' : 253-328-3
missing translation for 'tsca' : No
Recommended Storage : Store at -20°C
RUO – Research Use Only