missing translation for 'onlineSavingsMsg'
Learn More

Adenosine 5'-monophosphate sodium salt hydrate, 99%, Thermo Scientific Chemicals

Product Code. 11986851
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149022-20-8
Molecular Formula C10H13N5NaO7P
Molecular Weight (g/mol) 369.21
MDL Number MFCD18785650
InChI Key FYWLYWMEGHCZAX-GWKNMROSNA-M
Synonym sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate x:1:x, adenosine 5-monophosphoric acid disodium salt, adenosine 5'-monophosphate sodium salt, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate, pubchem14181, sodium 2r,3s,4r,5r-5-6-amino-9h-purin-9-yl-3,4-dihydroxytetrahydrofuran-2-yl methyl phosphate hydrate 2:1:x, 5'-adenylic acid,sodium salt, hydrate 9ci, disodium hydrate adenosine-5-monophosphate 2-
PubChem CID 23674992
IUPAC Name sodium [(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl hydrogen phosphate
SMILES [Na+].NC1=C2N=CN([C@@H]3O[C@H](COP(O)([O-])=O)[C@@H](O)[C@H]3O)C2=NC=N1

Specifications

Color White
CAS Min % 98.5
CAS Max % 100.0
pH 4.50 to 6.50
Physical Form Powder
Quantity 1 g
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 369.21
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Adenosine 5′-monophosphate sodium salt hydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'