missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adenosine 5'-triphosphate, disodium salt hydrate, 98%, Thermo Scientific Chemicals
152.76 € - 556.77 €
Chemical Identifiers
CAS | 34369-07-8 |
---|---|
Molecular Formula | C10H14N5Na2O13P3 |
Molecular Weight (g/mol) | 551.15 |
MDL Number | MFCD00150755 |
InChI Key | TTWYZDPBDWHJOR-WCYUCLFNNA-L |
PubChem CID | 131664345 |
IUPAC Name | disodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate |
SMILES | [Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10316070
|
Thermo Scientific Acros
102800100 |
10 g | Glass bottle |
152.76 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11936801
|
Thermo Scientific Acros
102800500 |
50 g | Glass bottle |
556.77 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
34369-07-8 | |
551.15 | |
TTWYZDPBDWHJOR-WCYUCLFNNA-L | |
disodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate |
C10H14N5Na2O13P3 | |
MFCD00150755 | |
131664345 | |
[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1 |
Specifications
188.0°C to 190.0°C | |
Brown to Colorless | |
100.0 | |
Liquid | |
C10H14N5Na2O13P3 | |
14,155 | |
(5% in water) Clear colorless | |
[Na+].[Na+].NC1=C2N=CN([C@@H]3O[C@H](COP([O-])(=O)OP([O-])(=O)OP(O)(O)=O)[C@@H](O)[C@H]3O)C2=NC=N1 | |
551.15 | |
551.14 | |
Authentic | |
Adenosine 5'-triphosphate, disodium salt hydrate |
34369-07-8 | |
97.5 | |
2.5 to 3.5 (5% aq. soln.) | |
97.5% min. (HPLC) | |
MFCD00150755 | |
atp disodium salt hydrate, atp disodium salt, atp disodium hydrate, adenosine 5'-triphosphate disodium salt hydrate, atp disodium salt trihydrate, 56-65-5 non-sodium, 987-65-5 anhydrous, 5 inverted exclamation marka-atp-na2, adenosine 5'-triphosphate disodium sal | |
TTWYZDPBDWHJOR-WCYUCLFNNA-L | |
disodium phosphono ({[(2R,3S,4R,5R)-5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphonato}oxy)phosphonate | |
131664345 | |
98% | |
(5% aq. soln.) clear, colorless |
RUO – Research Use Only