missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Adenosine-5'-triphosphate disodium salt hydrate, ultrapure, 98%
Click to view available options
Quantity:
25 g
100 g
Description
Adenosine 5'-Triphosphate, is used as a p2 purinergicagonist. It is also used as a nucleotide, the 5'-triphosphate of deoxyadenosine activated precursor in DNA synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Adenosine 5′-Triphosphate, is used as a p2 purinergicagonist. It is also used as a nucleotide, the 5′-triphosphate of deoxyadenosine activated precursor in DNA synthesis.
Solubility
Soluble at 5% (w/v) in water
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away fom strong oxidizing agents. Stable under ordinary conditions. Light sensitive, Hygroscopic
Adenosine 5′-Triphosphate, is used as a p2 purinergicagonist. It is also used as a nucleotide, the 5′-triphosphate of deoxyadenosine activated precursor in DNA synthesis.
Solubility
Soluble at 5% (w/v) in water
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away fom strong oxidizing agents. Stable under ordinary conditions. Light sensitive, Hygroscopic
Specifications
Specifications
Color | White |
Quantity | 100 g |
Sensitivity | Hygroscopic; Light sensitive |
Merck Index | 14,155 |
Solubility Information | Soluble at 5% (w/v) in water |
Formula Weight | 605.19 (551.15 Anhydrous) |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Adenosine-5'-triphosphate disodium salt hydrate |
Grade | Ultrapure |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'