Learn More
3M™ Adflo™ Powered Air Purifying Respirator System with 3M™ Speedglas™ G5-01 Welding Helmet
3M™ Adflo™ Powered Air Purifying Respirator System with 3M™ Speedglas™ G5-01 Series Welding Helmet provides all of the benefits of our G5-01 series welding helmet with our convenient Adflo™ Powered Air Purifying Respirator system.
Brand:  3M™ 7100257943
1792.90 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17895666
  • 2330.51 € / Each
Estimated Shipment: 29-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • For workers in heavy duty environments, 3M™ Adflo™ Powered Air Purifying Respirator with Speedglas™ G5-01 Series Heavy-Duty Welding Helmet system provides High-Efficiency particulate respiratory protection for welders in a robust yet compact design with ergonomic, adjustable head suspension.
  • Flip front welding shield allows for a clear, wide-view protective visor for grinding and setup.
  • The 3M™ Adflo™ Powered Air Respirator has a High-Efficiency particulate filter that can be stacked onto an optional 3M™ Adflo™ Filter (not included) for additional protection against certain vapours or a Nuisance Odour Pad Assembly.
  • Audible and visual alarms signal low airflow and low battery charge.
  • Available in two different Auto-Darkenig Lens options (VC/TW). G5 Series storage/carry bag included.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7 to 9 hr.
COMBINATION
G5-01VC
One Size
No
Lithium-ion
Black
PAPR System for Welding
PAPR, Welding Helmet