Learn More
3M™ Adflo™ Spark Arrestor, 836000
Replacement spark arrestor prevents sparks from entering the filer. For use with Adflo™ powered air respirator.
Brand: 3M™ 7000028010
10.99 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Check your spark arrestor regularly and assess for any damage
- Replace immediately if any damage is evident
- To increase service life, check equipment regularly
- Genuine 3M Accessory
Specifications
Black |