Learn More
ITW Chemische Produkte™ CleanStep™ Adhesive Cleanroom Mats
Reduce contamination in high traffic areas. ITW Chemische CleanStep™ Adhesive Cleanroom Mats are constructed with either 30 or 60 layers. Each removable sheet has a consistent adhesive coating designed to trap particles and prevent transfer.
Brand:  ITW Chemische Produkte™ AMA184682B
193.14 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
 View more versions of this product
Product Code. 11555325
  • 222.00 € / Pack of 4
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed to capture particulates from foot and wheel borne traffic prior to entry into cleanrooms or controlled environments
  • Provide a portable contamination control solution
  • Can be used with or without frames (sold separately)
  • Contain anti-microbial preservatives that prevent microbial growth
  • Consistent adhesive coating on each layer of 30 and 60 layer mats
  • Possible applications include cleanroom and critical environment entry points, gowning rooms or anterooms in the semiconductor, biotechnology, medical device, pharmaceutical processing, aerospace, and automotive markets
Specifications
Specifications
18 x 46 in. (46 x 117 cm)
60 layer
Blue
Polyethylene