missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Adhesive Multi-Purpose Polyester Labels

These Adhesive Multi-Purpose Polyester Labels allow for easy labeling of components with the thermal transfer, glossy permanent polyester label material.

Brand:  Brady™ 170735

Product Code. 17990700

  • 39.65 € / Pack of 280

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Versatile: Designed for a wide range of applications such as labeling equipment, circuit boards, panel components and barcodes
  • Tough: Constructed to withstand abrasions, extreme temperatures and solvents such as alcohol, oil and water
  • Adhesive: Made with a permanent acrylic adhesive that sticks to rough, smooth and powder-coated flat surfaces
  • Certified: UL Recognized to UL969, CSA to C22.2 No.0.15-95, RoHS Compliant to 2011/65/EU (Pass 2015/863)
  • Print almost anywhere with a durable Brady handheld label printer
  • For material specific performance and properties, please refer to the Technical Data Sheet (TDS)
Specifications

Specifications

M4-7-422
Black on White
Cartridge (Tape)
Labels
Asset Tracking, Barcode Labeling, Circuit Board Labeling, Component and Equipment Labeling, Data and Telecommunications Labeling, General Identification, Inventory and Inspection Labeling
280 Pk.
0.004 in.
0.5 x 0.5 in.
0.5 in.
Permanent Acrylic
Gloss
Polyester
Non-sterile
BMP41, BMP51, BMP53, M511
-40°F to +212°F
0.102 mm
0.5 in.
12.70 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.