missing translation for 'onlineSavingsMsg'
Learn More

Adipic dihydrazide, 98%, Thermo Scientific Chemicals

54.77 € - 609.03 €

Chemical Identifiers

CAS 1071-93-8
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00007614
InChI Key IBVAQQYNSHJXBV-UHFFFAOYSA-N
Synonym adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m
PubChem CID 66117
IUPAC Name hexanedihydrazide
SMILES NNC(=O)CCCCC(=O)NN
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10531651
View Documents
Thermo Scientific Acros
203310250
25 g Plastic bottle
54.77 €
25g
Please to purchase this item. Need a web account? Register with us today!
10016023
View Documents
Thermo Scientific Acros
203311000
100 g Plastic bottle
130.65 €
100g
Please to purchase this item. Need a web account? Register with us today!
10076063
View Documents
Thermo Scientific Acros
203315000
500 g Plastic bottle
609.03 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1071-93-8
174.20
IBVAQQYNSHJXBV-UHFFFAOYSA-N
66117
NNC(=O)CCCCC(=O)NN
C6H14N4O2
MFCD00007614
adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m
hexanedihydrazide

Specifications

1071-93-8
100.0
White to Yellow
98%
H2NNHCO(CH2)4CONHNH2
02, I, 277
Solubility in water: soluble. Other solubilities: soluble in acetic acid,slightly soluble in acetone,insoluble in ethanol,ether and benzene
NNC(=O)CCCCC(=O)NN
174.20
174.2
Crystalline Powder
97.5
178.0°C to 182.0°C
Authentic
C6H14N4O2
MFCD00007614
adipic dihydrazide, adipohydrazide, adipic acid dihydrazide, hexanedioic acid, dihydrazide, adipyl hydrazide, hexanediohydrazide, adipic acid, dihydrazide, adipodihydrazide, hexanedioic acid, 1,6-dihydrazide, unii-vk98i9yw5m
IBVAQQYNSHJXBV-UHFFFAOYSA-N
hexanedihydrazide
66117
98%
Adipic dihydrazide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 213-999-5

Product Identifier
  • Adipic dihydrazide

Signal Word
  • Warning

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Respiratory sensitiser Category 1

Hazard Statement
  • H411-Toxic to aquatic life with long lasting effects.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.