missing translation for 'onlineSavingsMsg'
Learn More
Adipoyl chloride, 98%, Thermo Scientific Chemicals Product Code.: 10216040

Adipoyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 10216040
500 mL, Glass bottle
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10216040

Brand: Thermo Scientific Acros 173185000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Adipoyl chloride, 98%, C6H8Cl2O2, CAS Number-111-50-2, adipic acid chloride, adipic acid dichloride, adipic dichloride, adipoylchloride, adipoyl chloride, adipoyl dichloride, adipyl chloride, butane-1,4-dicarbonyl chloride, hexanedioyl chloride, pubchem13242, 500mL, 02, 653

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-50-2
Molecular Formula C6H8Cl2O2
Molecular Weight (g/mol) 183.03
MDL Number MFCD00000759
InChI Key PWAXUOGZOSVGBO-UHFFFAOYSA-N
Synonym adipoyl chloride, adipoyl dichloride, adipyl chloride, adipoylchloride, adipic dichloride, hexanedioyl chloride, adipic acid dichloride, butane-1,4-dicarbonyl chloride, adipic acid chloride, pubchem13242
PubChem CID 61034
IUPAC Name hexanedioyl dichloride
SMILES C(CCC(=O)Cl)CC(=O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown
Density 1.2500g/mL
Boiling Point 105.0°C to 107.0°C (2.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClCO(CH2)4COCl
Refractive Index 1.4700 to 1.4720
Quantity 500 mL
Beilstein 02, 653
Specific Gravity 1.25
Solubility Information Solubility in water: reacts. Other solubilities: forming hcl
Formula Weight 183.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Adipoyl chloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Adipoyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Adipoyl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.