missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OHAUS™ Adjustment Bar
OHAUS™ Adjustment Bar is an accessory for use with adjustable platforms, which are designed to be mounted onto heavy duty shakers.
Brand: OHAUS™ 30400073
62.60 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with platforms, which increase the amount of usable space for samples
- Heavy duty shaker accessory
Specifications
Adjustment Bar | |
Black | |
Heavy Duty Shakers |
460 mm | |
Adjustment Bar, 46 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction