missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Adonitol, 98%

Product Code. 10657332
Click to view available options
Quantity:
5 g
10 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Adonitol, 98%, C5H12O5, CAS Number-488-81-3, adonit, adonite, adonitol, d-ribitol, d-xylitol, eutrit, ribitol, xylit, xylite, xylitol, 100g, 97.5% min. (ELSD) (HPLC), 01,53, 100.0, 99.0, White, 207-685-7, 152.15, HEBKCHPVOIAQTA-NGQZWQHPSA-N, Authentic, (2S,4R)-pentane-1,2,3,4,5-pentol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 488-81-3
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291
InChI Key HEBKCHPVOIAQTA-NGQZWQHPSA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES C(C(C(C(CO)O)O)O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 101.0°C to 104.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 01,53
Merck Index 15,157
Solubility Information Solubility in water: soluble. Other solubilities: soluble in hot alcohol,insoluble in ether
Formula Weight 152.15
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Adonitol
Show More Show Less
Product Identifier
  • Adonitol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.