Learn More
Grant Instruments™ Advanced Air Cleaner Recirculator, 2 Lamps
Powerful/efficient UV cleaner-recirculator. Two UV lamps providing air disinfection in hospitals, research laboratories and clinics. The UV air flow cleaner-recirculator consists of 2 germicidal UV lamps, a fan, unit equipped with dust filters and a control unit, confined in a flow through chamber.
Marca: Grant Instruments™ UVR-Mi EU
Descripción
The inner mirror surface of the recirculator chamber reflects ultraviolet rays thereby increasing the density of UV radiation and enhancing the disinfection effect.
- Two UV-lamps
- Full user protection from direct UV radiation;
- Two operation modes — short time (under timer control) and continuous operation
- Digital control unit allows tracking of overall UV lamp operating time
- 1 min-24 hours/non stop timer
Recirculators fixation:
- Convenient fixation on walls (standard)
- Mounting on a movable tripod (optional)
Especificaciones
18 mW/cm2/sec, 14 m3/hour | |
2 Lamps | |
UVR-S | |
UV-Air Flow Cleaner-Recirculator |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.