Learn More
Grant Instruments™ TX150 Series Heated Circulating Bath, Stainless Steel Tank
Provides outstanding performance for routine and sophisticated applications requiring accurate temperature control up to a maximum of 150°C. Grant Instruments™ Optima™ TX150 Series Digital Heated Circulating Bath, Stainless Steel Tank combines the powerful and versatile Grant Optima™ TX150 digital heating circulator with a choice of five stainless steel (ST) tanks with different capacities.
Brand:  Grant Instruments™ TX150-ST5
 View more versions of this product
Product Code. 12737839
  • 2023.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Series of digitally controlled heated circulating baths provides outstanding performance for routine and sophisticated applications requiring accurate temperature control up to a maximum of 150ºC.
  • Cmbine the powerful and versatile Grant Optima™ TX150 digital heating circulator with a choice of three economy plastic (P) tanks or five stainless steel (ST) tanks with different capacities
  • Heating circulator incorporates Intelligent Control Optimisation™ (ICO) for adaptive intelligent PID temperature control
  • Includes a powerful integral pump, making TX150 systems suitable both for immersing samples and circulating temperature controlled fluid to external devices 
Specifications
Specifications
0°C to 200°C
Stainless Steel
150 mm
Includes TX150 immersion heater/stirrer with ST5 Bath
150 mm
300 mm