Learn More
Grant Instruments™ TX150 Series Heated Circulating Bath, Stainless Steel Tank
Provides outstanding performance for routine and sophisticated applications requiring accurate temperature control up to a maximum of 150°C. Grant Instruments™ Optima™ TX150 Series Digital Heated Circulating Bath, Stainless Steel Tank combines the powerful and versatile Grant Optima™ TX150 digital heating circulator with a choice of five stainless steel (ST) tanks with different capacities.
Brand:  Grant Instruments™ TX150-ST18
 View more versions of this product
Product Code. 12757839
  • 2445.00 € / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Stability ±0.01ºC
  • Large, bright full colour display
  • Icon driven home screen via a dial and two function buttons
  • Programming/temperature profiling (TX150, 1 program with 30 segments)
  • Model available with/without clamp (T-Clamp)
  • Low-liquid detection (float switch)
  • 5 point user calibration
  • User adjustable over temperature dial
  • 5 point user calibration
Specifications
Specifications
0°C to 200°C
Stainless Steel
150 mm
Includes TX150 immersion heater/stirrer with ST18 Bath
300 mm
505 mm