Learn More
OHAUS™ Adventurer™ Precision Balance
Intuitive Adventurer™ Precision Balance designed for routine weighing.
Brand:  OHAUS™ 30805891
2190.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17991292
  • 2245.00 € / 1 unit
Explore more special offers
Description
Show More
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
OHAUS™ Adventurer™ Precision Balance > Model: AX623/E, Capacity: 620.0 g, Readbility: 0.001 g, Calibration: External