Learn More
Motic™ AE31E Binocular Microscope
Use this microscope for routine live cell inspection. Motic™ AE31E Inverted Binocular Microscope is ideal for university, routine-clinical and lab environments.
Brand:  Motic™ 1100100200565
Product Code. 15358805
  • 3925.00 € / Each
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Centering telescope.
  • CCIS Plan achromatic objective PL4X
  • CCIS Plan achromatic Phase objective PL PH10X, LWD PL PH20X, LWD PL PH40X
  • Plain stage, glass and metal inserts
  • Phase slider, centerable PH1, brightfield, PH3
Specifications
Specifications
100/240 V
Side-facing quintuple nosepiece.
Widefield High Eyepoint WFPL10X/22 mm with diopter adjustment on both eyepieces and rubber eyecups
Inverted Binocular Microscope
ELWD N.A. 0.3 with phase slider (Ph1, BF)
Universal power supply 100/240 V (CE)
Quartz Halogen
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Motic™ AE31E Binocular Microscope > Product Type: Inverted Binocular Microscope