missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AEBP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540406
This item is not returnable.
View return policy
Description
Peptide sequence: VPPEKTKDKG KKGKKDKGPK VPKESLEGSP RPPKKGKEKP PKATKKPKEK Sequence homology: Cow: 92%; Dog: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 92%; Rabbit: 100%; Rat: 100%
AEBP1 Polyclonal antibody specifically detects AEBP1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
AEBP1 | |
Polyclonal | |
Unconjugated | |
AEBP1 | |
ACLP; adipocyte enhancer-binding protein 1; AE binding protein 1; AE-binding protein 1; AEBP1; Aortic carboxypeptidase-like protein; FLJ33612; transcription factor AEBP1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
165 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8IUX7 | |
AEBP1 | |
Synthetic peptide directed towards the N-terminal of human AEBP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction