Learn More
Tocris Bioscience™ AEBSF
Broad spectrum, irreversible inhibitor of serine proteases
Brand:  Tocris Bioscience™ 5175/100
Product Code. 18705698
  • 166.00 € / 100mg
Explore more special offers
Description
Description
AEBSF is a broad spectrum, irreversible inhibitor of serine proteases. Also inhibits NADPH oxidase activation in vitro. Attenuates airway inflammation in a mouse model of airway allergy.
Specifications
Specifications
>98%
AEBSF
AEBSF
100 mg
239.69