Learn More
Eppendorf™ Aerosol-Tight Cap, Universal Buckets, S-4xUniversal Rotor
Seals centrifuge rotor buckets for protection against hazardous materials.
Brand: Eppendorf™ 5910750005
223.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Suitable for S-4xUniversal Rotor buckets
- Creates aerosol-tight seal when working with hazardous substances
Specifications
S-4xUniversal rotor buckets |