Learn More
Eppendorf™ Aerosol-tight caps
Use these caps with S-4x1000 plate/tube bucket, S-4x750 plate bucket or S-4x Universal Large Rotor
Brand: Eppendorf™ 5895111009
223.00 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Set of 2 pcs.Specifications
S-4x1000 plate/tube bucket and S-4x750 plate bucket |