missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Aerosol-Tight Rotor Lids

Providing secure closure for a variety of Eppendorf rotors.

Brand:  Eppendorf™ 5810724007

189.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10354902

  • 199.00 € / Pack of 2

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Aluminim construction
  • Aerosol-tight
  • Autoclavable for 20 min at 121°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Aerosol-tight Lid
Aluminum
Rotor A-4-81
Autoclavable (121°C for 20 min.)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers