missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human AFP ELISA Kit

ELISA

678.00 € - 2780.00 €

Specifications

Accession Number P02771
Assay Range 7.37 to 1800 pg/mL
Assay Sensitivity 6 pg/mL
Calibration Range 6 pg/mL to 1800 pg/mL
Assay Solid-phase sandwich ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15414893
View Documents
Invitrogen™
EHAFP
96 Tests
678.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
17165339
View Documents
Invitrogen™
EHAFPX5
5 x 96 Tests
2780.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The Human Alpha FetoProtein (Hu AFP) ELISA quantitates Hu AFP in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu AFP. Principle of the method The Human AFP solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

AFP (Alpha Fetoprotein) is a major plasma protein produced by the yolk sac, intestinal tract and the liver during fetal life. AFP expression in adults is often associated with hepatoma or teratoma. However, hereditary persistence of AFP may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and AFP and albumin genes are present in tandem in the same transcriptional orientation on chromosome 4. AFP is found in monomeric, dimeric, and trimeric forms. AFP can also bind to copper ions, nickel ions, fatty acids and bilirubin. The level of AFP in amniotic fluid is used to measure renal loss of protein to screen for spina bifida and anencephaly. High level of serum AFP has been identified in patients with hepatocellular carcinomas (HCC), teratoblastoma, colorectal cancer, pancreatic cancer, and germ cell neoplasms.
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Human AFP ELISA Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.