Learn More
Thermo Scientific Chemicals Agar powder
45.43 € - 593.96 €
Chemical Identifiers
CAS | 9002-18-0 |
---|---|
Molecular Formula | C14H24O9 |
Molecular Weight (g/mol) | 336.337 |
MDL Number | MFCD00081288 |
InChI Key | GYYDPBCUIJTIBM-DYOGSRDZSA-N |
Synonym | agar, agar, pure, powder, agar agar bacteriological, 3r,4s,5s,6r-2-4r,5s-4-hydroxy-3-methyl-2,6-dioxabicyclo 3.2.1 octan-8-yl oxy-6-hydroxymethyl-4-methoxyoxane-3,5-diol |
PubChem CID | 71571511 |
IUPAC Name | (2R,3S,4S,5R)-2-(hydroxymethyl)-6-[[(4R,5S)-4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-methoxyoxane-3,5-diol |
SMILES | CC1C(C2C(C(O1)CO2)OC3C(C(C(C(O3)CO)O)OC)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11479632
|
Thermo Scientific Chemicals
A10752.22 |
100 g |
45.43 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11489632
|
Thermo Scientific Chemicals
A10752.36 |
500 g |
181.91 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499632
|
Thermo Scientific Chemicals
A10752.0E |
2500 g |
593.96 €
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Agar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAgar is the most suited solidifying agent for scientific applications that require incubation temperatures closer to human body temperature. Agar is used as a medium in bacteriology and plant biology, impression material in dentistry, biofertilizer in organic farming, and salt bridges in electrochemistry. It can be used to measure the motility and mobility of microorganism owing to the porous nature of the gel form of an agar or agarose medium.
Solubility
Soluble in water at temperatures between 950C and 1000 C. Insoluble in cold water.
Notes
Prolonged exposure to high temperatures can degrade Agar powder.
Chemical Identifiers
9002-18-0 | |
336.337 | |
GYYDPBCUIJTIBM-DYOGSRDZSA-N | |
71571511 | |
CC1C(C2C(C(O1)CO2)OC3C(C(C(C(O3)CO)O)OC)O)O |
C14H24O9 | |
MFCD00081288 | |
agar, agar, pure, powder, agar agar bacteriological, 3r,4s,5s,6r-2-4r,5s-4-hydroxy-3-methyl-2,6-dioxabicyclo 3.2.1 octan-8-yl oxy-6-hydroxymethyl-4-methoxyoxane-3,5-diol | |
(2R,3S,4S,5R)-2-(hydroxymethyl)-6-[[(4R,5S)-4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-methoxyoxane-3,5-diol |
Specifications
Powder | |
C14H24O9 | |
agar, agar, pure, powder, agar agar bacteriological, 3r,4s,5s,6r-2-4r,5s-4-hydroxy-3-methyl-2,6-dioxabicyclo 3.2.1 octan-8-yl oxy-6-hydroxymethyl-4-methoxyoxane-3,5-diol | |
GYYDPBCUIJTIBM-DYOGSRDZSA-N | |
(2R,3S,4S,5R)-2-(hydroxymethyl)-6-[[(4R,5S)-4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-methoxyoxane-3,5-diol | |
71571511 |
9002-18-0 | |
MFCD00081288 | |
Soluble in water at temperatures between 950C and 1000C. Insoluble in cold water. | |
CC1C(C2C(C(O1)CO2)OC3C(C(C(C(O3)CO)O)OC)O)O | |
336.337 | |
Agar |
Safety and Handling
missing translation for 'einecsNumber' : 232-658-1
missing translation for 'rtecsNumber' : AW7950000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.