missing translation for 'onlineSavingsMsg'
Learn More
Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™ Product Code.: 10776644

Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™

Product Code. 10776644
500 g, Poly Bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 10776644

Brand: Fisher BioReagents BP1356500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C12H18O9

Specifications

Physical Form Powder/Solid
Color Cream
Quantity 500 g
DNase DNase free
Electroendosmosis (EEO) 0.15 max.
Gelation Temperature 34.5° to 37.5°C
Gel Strength 1200g/cm2min.
Packaging Poly Bottle
Formula Weight 306.12
Grade DNase- and RNase-Free
Chemical Name or Material Agarose, Broad Separation Range for DNA/RNA (500bp to 25kb)
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Agarose (Broad Separation Range for DNA/RNA/Genetic Analysis Grade), Fisher BioReagents™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.