missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Agarose Gel Loading Dye (6X, Glycerol-Based, no Xylene Cyanol FF)

67.84 € - 97.38 €

Specifications

Chemical Name or Material Agarose gel loading dye
Physical Form Liquid
Name Note glycerol based, Xylene Cyanol FF free
Concentration 6X
Solubility Information Fully misicible in water.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15480157
View Documents
Thermo Scientific Chemicals
J60933.AC
25 mL
67.84 €
25mL
In Stock
to see stock.
Add to basket
 
15490157
View Documents
Thermo Scientific Chemicals
J60933.AD
50 mL
97.38 €
50mL
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
 
Description

Description

6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6X DNA Loading Buffer is a pre-mixed solution used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is a pre-mixed loading buffer containing two different dyes xylene cyanol FF for visual tracking of DNA migration during agarose gel electrophoresis.

Solubility
Fully misicible in water.

Notes
No Xylene cyanol FF. Contains 0.25% bromophenol blue, 30% glycerol.
Specifications

Specifications

Agarose gel loading dye
glycerol based, Xylene Cyanol FF free
Fully misicible in water.
Liquid
6X
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
If eye irritation persists: Get medical advice/attention.

Warning

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.