missing translation for 'onlineSavingsMsg'
Learn More

Melford Agarose Gel Loading Dye

6X Concentrated gel loading dye for molecular biology applications.

Brand:  Melford A21500-25.0

Product Code. 30018017

  • 85.70 € / 25mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

6X Concentrated gel loading dye for molecular biology applications.

6X Concentrated gel loading dye for molecular biology applications.

Formulation:

Bromophenol Blue: 0.25%

Xylene Cyanol FF: 0.25%

Glycerol: 40% in sterile water

Specifications

Specifications

25 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers